Learn More
Cytiva Supor EKV membrane in Mini Kleenpak™ capsules, 0.2 µm removal rating, 200 cm² EFA, ¼-½ in. hose barb connections, suitable for gamma irradiation, box of 3
Supplier:  Cytiva KA02EKVP2G
Encompass
Capsule Filtration of Buffers, Tissue Culture Media, and Other Biological Fluids
Catalog No. NC9960246