Learn More
Chemglass Life Sciences Filter Funnel, Disposable, 40-60mL, 10 Micron Polyethylene Frit, Flared Top
SDP
Supplier:  Chemglass Life Sciences OP660212
Encompass_Preferred
Filter Funnel, Disposable, 40-60mL, 10 Micron Polyethylene Frit. This unique disposable filter funnel system for solid-liquid separation can be used in chemistry and biological laboratories. The system consists of a polypropylene filter funnel with a 10 micron polyethylene fritted disc, a CG-1049-J style glass vacuum adapter, and a flask or vial. The CG-1049-J vacuum adapters are specially designed so that the vacuum can be controlled using a PTFE stopcock to prevent the filtrates from being sucked into the vacuum line. Also, the disposable filter funnels have a 80mm long stem so that the glass vacuum adapter is not contaminated during the filtration process. U.S. Patent Pending
Catalog No. 50-121-4687