Learn More
Hach Company TSS Glass Fiber Filter, Pore Size 1.5 µm, Diameter 20 mm, 100/pk
Supplier:  Hach Company 2198720
Encompass
TSS Filters are recommended for use in Standard Methods 2540 C and 2540 D. Optimized surface characteristics of this binderless borosilicate glass microfiber filter eliminates pin-holing and fiber-shedding to ensure consistent, accurate results comparable to 934-AH brand filters. Fast flow rate, fine porosity and 1.5 μm particle retention makes the TSS Filter the preferred choice of environmental chemists for total suspended solids analysis of water and wastewater.934-AH is a registered trademark of Whatman, Inc./GE Healthcare.
Catalog No. 50-199-5549