Learn More
Waters Corp Alliance Filter Insert Assembly, 2μm, Stainless Steel, HPLC System
Supplier:  Waters Corp WAT088084
Encompass_Preferred
  • Extends column life by capturing particulates and debris, preventing clogging and damage to analytical columns.
  • Enhances analytical performance, reducing baseline noise and peak broadening for sharper peaks and improved resolution.
  • Seamless compatibility: designed to integrate with HPLC systems without significant modifications.
  • Efficient operation under high-pressure HPLC conditions, delivering superior filtration without impeding flow rates.
  • Durable construction ensures consistent performance under rigorous laboratory conditions.
  • Easy replacement minimizes downtime and maintains productivity.
Catalog No. 50-819-112