Learn More
Hach Company Filter, Membrane, Cellulose, Pore Size 0.8 µm, Diameter 47 mm, 100/pk, plain, non-sterile
Supplier:  Hach Company 2640800
Encompass
Pall GN-4 Metricel membrane filters are made of mixed cellulose esters. Non-sterile, 0.8µm Disc Filters are suitable for air monitoring applications and meets NIOSH requirements for airborne metals and asbestos monitoring. Dissolves completely using standard digestion procedures, and clears completely for minimal interference in fiber counting. Pall GN Metricel Membrane Disc Filters autoclavable.
Catalog No. 50-199-6433