Learn More
Sterlitech Corporation Polycarbonate (PCTE) Membrane Filters, 0.4 Micron, 47mm, 100/Pk
SDP
Supplier:  Sterlitech Corporation PCT0447100
Encompass_Preferred
Polycarbonate Track Etch (PCTE) membranes are made from a thin, microporous polycarbonate film material. It is ideally suited for use in blood assays and high-purity and general filtration.
Catalog No. NC1543745