Learn More
BD Cell Analysis 3P BD Biosciences Quick Conn Filter, 0.2 um Pore Size, Sterile-Grade, Unit Each
Supplier:  BD Cell Analysis 3P 661744
Encompass
The BD Biosciences Filter Quick Conn FEM FEM 0.2um PPL is a sterile sheath fluid filter optimized for flow cytometry systems, ensuring purity by removing particulates and microorganisms through its 0.2 um pore size. Its female-to-female quick connections allow for easy installation, enhancing workflow efficiency in research labs. Designed explicitly for research use only, it supports the integrity of research samples.
Catalog No. NC1308229