Learn More
Grainger Air Filter Roll, 45 ft. Nominal Width, 10" Nominal Height, MERV 8, Polyester
Supplier:  Grainger 2GGK1
Encompass_Preferred
General use air filter rolls capture airborne contaminants to protect HVAC systems from damage. They consist of a long length of filter media that can be cut to size. These rolls are most commonly used as pre-filters to capture large particles before they reach the final filters downstream, which extends the life of the more expensive final filters.
Catalog No. NC1692135