Learn More
Chemglass Life Sciences Filter Tube, 24/40 Inner Joints, 40mm OD Medium Frit, Airfree, Schlenk
SDP
Supplier:  Chemglass Life Sciences AF054408
Encompass_Preferred
Filter Tube, 24/40 Inner Joints, 40mm OD Medium Frit, Airfree, Schlenk. Tube allows for filtration followed by collection of crystals on opposite side of fritted disc. The standard taper inner joints at both ends are the same size. Available with a choice of coarse or medium porosity fritted discs.
Catalog No. AF054408