Learn More
IDEX Health & Science Upchurch FILTER WATERS SOLVENT INLET
Supplier:  IDEX Health & Science Upchurch A310
Encompass_Preferred
Idex Inlet Filter, Stainless Steel, Pore Size: 10 um, Inlet Solvent Filter, Flow Rate: 40 mL/min., For Use With: To filter out particulate matter from the solvent that may otherwise damage hardware, for general use, preparative, specialized applications, Waters analytical HPLC systems
Catalog No. NC1920933