Learn More
Microsolv Technology Corporation Filters Precolumn Ss 2Um 10/Pk
Supplier:  Microsolv Technology Corporation 49650020
Encompass_Preferred
Inline Filters, for HPLC Columns, 2.0um Stainless Steel Filter Element and all PEEK body. Compatible with all Analytical HPLC Columns. Disposable, with End Plugs. Natural, MicroSolv Brand. 10/PK.
Catalog No. 50-014-52550