Learn More
Medchemexpress LLC FIMEPINOSTAT 10MG
SDP
Supplier:  Medchemexpress LLC HY1352210MG
Encompass_Preferred
Fimepinostat (Synonyms: CUDC-907.)Fimepinostat (CUDC-907) potently inhibits class I PI3Ks as well as classes I and II HDAC enzymes with an IC50 of 19/54/39 nM and 1.7/5.0/1.8/2.8 nM for PI3Kα/PI3Kβ/PI3Kδ and HDAC1/HDAC2/HDAC3/HDAC10 , respectively.CAS 1339928-25-4
Catalog No. NC1783205