Learn More
Selleck Chemical LLC Fimepinostat5mg
SDP
Supplier:  Selleck Chemical LLC S27595MG
Encompass_Preferred
CUDC-907 is a dual PI3K and HDAC inhibitor for PI3K? and HDAC1/2/3/10 with IC50 of 19 nM and 1.7 nM/5 nM/1.8 nM/2.8 nM, respectively. CUDC-907 induces cell cycle arrest and apoptosis in breast cancer cells. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3103