Learn More
Enzo Life Sciences Finasteride (500mg). CAS: 98319-26-7
SDP
Supplier:  Enzo Life Sciences ALX270491M500
Encompass_Preferred
Inhibitor of 5α-reductase, the enzyme which converts testosterone to the more potent androgen, 5α-dihydrotestosterone. Used as a treatment in benign prostatic hyperplasia and prostate cancer. Also used to treat male pattern hair loss. Alternative name: MK-906. Purity: ≥98% (Assay (UV)). Appearance: White to off-white crystalline powder. Solubility: Soluble in DMSO, 100% ethanol, methanol or propanol. Long Term Storage: +4°C.
Catalog No. 50-200-8617