Learn More
Medchemexpress LLC Finasteride | 98319-26-7 | 372.54 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13635100MG
Encompass_Preferred
Finasteride, also known as MK-906, is an orally active and competitive 5α-reductase inhibitor. It exhibits an IC50 of 4.2 nM for type II 5α-reductase, demonstrating approximately a 100-fold greater affinity for the type II enzyme compared to the type I enzyme.
  • Utilized for research related to benign prostatic hyperplasia (BPH)
  • Applied in studies concerning androgenic alopecia
Catalog No. 50-003-50248