Learn More
Medchemexpress LLC Finasteride | 98319-26-7 | MFCD00869737 | 100.0% | 372.54 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY13635200MG
Encompass_Preferred
Finasteride is an orally active and competitive 5α-reductase inhibitor primarily targeting type II 5α-reductase. It works by blocking the conversion of testosterone into dihydrotestosterone (DHT), thereby reducing DHT levels in prostate cells and scalp hair follicles. This compound is used in research for conditions such as benign prostatic hyperplasia (BPH) and androgenic alopecia.
  • Orally active and competitive 5α-reductase inhibitor
  • Exhibits high affinity for type II 5α-reductase enzyme
  • Reduces prostatic size and increases urine flow rate
  • Promotes hair growth and prevents hair loss
  • Suitable for research on benign prostatic hyperplasia (BPH)
  • Suitable for research on androgenic alopecia
  • Suppresses DHT levels in plasma and prostate
Catalog No. 50-003-51147