Learn More
World Precision Instrument Fine Mesh Baskets with Lids-WP-4354
SDP
Supplier:  World Precision Instrument WP4354
Encompass
  • Manufactured with stainless steel 304
  • Electropolished
  • Burr free to protect wraps and prevent injury
  • Smooth rounded corners
  • Whole and wires size : 1.6x1.6x0.7 mm
Catalog No. 50-209-0217