Learn More
Fine Science Tools Fine Scissors, Sharp-Blunt, Stainless Steel, 10 cm
SDP
Supplier:  Fine Science Tools 1402910
Encompass
Tip Shape: Curved/ Tips: Sharp-Blunt/ Cutting Edge: 27mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No
Catalog No. NC1574927