Learn More
ATCC Neisseria elongata; subsp. glycolytica
Supplier:  ATCC 29315
Encompass
Neisseria elongata subsp. glycolytica strain 6171/75 [NCTC 11050] is a whole-genome sequenced bacterial type strain that was isolated from a throat swab. This strain is grown under a 5% CO2 atmosphere on GC medium.
Catalog No. 50-238-0597