Learn More
Medchemexpress LLC HY-111372 10mg , Finerenone CAS:1050477-31-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11137210MG
Encompass_Preferred
HY-111372 10mg Finerenone CAS: 1050477-31-0 Finerenone (BAY 94-8862) is a third-generation, selective, and orally available nonsteroidal mineralocorticoid receptor (MR) antagonist (IC50=18 nM). Finerenone displays excellent selectivity versus glucocorticoid receptor (GR), androgen receptor (AR), and progesterone receptor (>500-fold). Finerenone has the potential for cardiorenal diseases research, such as type 2 diabetes mellitus and chronic kidney disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0529