Learn More
Selleck Chemical LLC Finerenone S9702-25MG
SDP
Supplier:  Selleck Chemical LLC S970225MG
Encompass_Preferred
Finerenone (FIN BAY 94-8862) is a highly selective and orally available nonsteroidal antagonist of mineralocorticoid receptor (MR) with IC50 of 18 nM Finerenone has the potential for cardiorenal diseases research such as type 2 diabetes mellitus and chronic kidney disease
Catalog No. 50-000-24951