Learn More
Selleck Chemical LLC Fingolimod (FTY720) HCl S5002-1g
SDP
Supplier:  Selleck Chemical LLC S50021G
Encompass_Preferred
Fingolimod (FTY720 Fingolimod Hydrochloride) HCl is a S1P antagonist with IC50 of 0 033 nM in K562 and NK cells Please use saline solution rather than PBS for dilutions PBS may cause precipitation
Catalog No. 50-311-1735