Learn More
Cayman Chemical Fingolimod, 162359-55-9, 5 mg
SDP
Supplier:  Cayman Chemical 119755
Encompass_Preferred
Molecular Formula C19H33NO2, Purity ≥98%, Formula Weight 307.5, Synonyms: 2-amino-2-[2-(4-octylphenyl)ethyl]-1,3-propanediol; A fungal metabolite with immune modulating activities; prolongs allograft transplant survival in numerous models by inhibiting lymphocyte emigration from lymphoid organs; acts as an agonist at S1P receptors when phosphorylated; enhances activity of Abcb1 and Abcc1 and inhibits cytosolic PLA2 activity.
Catalog No. NC0517641