Learn More
Medchemexpress LLC Fingolimod phosphate | 402615-91-2 | 99.2% | 387.45 g/mol | C19H34NO5P | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1538110MG
Encompass_Preferred
Fingolimod phosphate is the phosphorylated, active metabolite of fingolimod (FTY720) that acts as a sphingosine-1-phosphate (S1P) receptor agonist. It is supplied as a research reagent for studies of immune modulation and neurological disease models. The compound is provided as a solid with recommended storage conditions and defined stability in solution.
  • Active metabolite used in S1P receptor and multiple sclerosis research.
  • High purity suitable for research applications.
  • Molecular formula C19H34NO5P; molecular weight 387.45 g/mol.
  • Offered in multiple package sizes, including 10 mg.
  • Solid storage: 4°C, protect from light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-87023