Learn More
Medchemexpress LLC Fingolimod phosphate | 402615-91-2 | C19H34NO5P | 1 MG
SDP
Supplier:  Medchemexpress LLC HY153811MG
Encompass_Preferred
Fingolimod phosphate is an orally active sphingosine 1-phosphate (S1P) receptor agonist. It promotes the neuroprotective effects of microglia and is used for research on multiple sclerosis and neurological diseases.
  • Orally active S1P receptor agonist
  • Promotes neuroprotective effects of microglia
  • Used for research on multiple sclerosis
  • Used for research on neurological diseases
  • Prevents lymphocytes from moving out of lymphoid organs
  • Inhibits autoreactive lymphocytes from infiltrating the central nervous system
  • Downregulates pro-inflammatory cytokines in activated microglia
  • Upregulates brain-derived neurotrophic factor and glial cell-derived neurotrophic factor production in microglia
Catalog No. 50-003-76031