Learn More
Hach Company Finished Mounting Hardware
Supplier:  Hach Company MH462G
Encompass
Each immersion hardware includes a 1/2-inch diameter x 4 foot long pipe, 1/2 x 3/4-inch NPT coupling, and plastic pipe-mount junction box with terminal strip. Used for 3700-series and PC/RC series sensors.
Catalog No. 50-199-8051