Learn More
Pipette Supplies, Inc. Finnpipette Novus Lithium-Ion Battery
SDP
Supplier:  Pipette Supplies, Inc. TS 2214980
Encompass
OEM Rechargeable Lithium-Ion Battery for the Finnpipette Novus Single Channel and Multichannel pipettes. Battery includes battery cover. 3.7V, 950mAh, 3.52Wh
Catalog No. NC3864799