Learn More
TARGETMOL CHEMICALS INC FINROZOLE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T273205MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Finrozole (MPV 2213ad) is a novel selective aromatase inhibitor that is partially reversible for breast development. purity: 100%
Catalog No. 50-258-0626