Learn More
AdipoGen Fipronil (100 mg)
SDP
Supplier:  AdipoGen AGCR13648M100
Encompass_Preferred
Chemical. CAS: 120068-37-3. Formula: C12H4Cl2F6N4OS. MW: 437.2. Alzheimer inducing agent (Alzheimerogen). Potent GABA receptor alpha-4 subunit and beta-3 subunit inhibitor. Broad-range insecticide. Blocks GABA A-gated chloride channels and glutamate-activated chloride channels (GluCls) in the central nervous system, resulting in overexcitation of the insects nervous system and muscles. Induces oxidative stress in mouse kidney and brain. Increases CYP1A1, CYP2B6, CYP3A4 and CYP3A5 transcript, protein and enzyme activity levels.
Catalog No. 50-205-4022