Learn More
Medchemexpress LLC Fipronil | 120068-37-3 | 99.5% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB082210MM/1ML
Encompass_Preferred
Fipronil is a broad-spectrum insecticide effective against Lepidoptera species, thrips, locusts, ants, cockroaches, fleas, and ticks. It selectively inhibits GABA receptors with IC50s of 30 nM and 1600 nM for cockroach and rat GABA receptors, respectively. It also targets glutamate-gated chloride channels (GluCls) in cockroaches and induces apoptosis in HepG2 cells, promoting the expression of CYP1A1 and CYP3A4 mRNA in human hepatocytes.
  • Broad-spectrum insecticide
  • Selectively inhibits GABA receptors
  • Targets glutamate-gated chloride channels (GluCls) in insects, which are not present in mammals
  • Induces apoptosis in HepG2 cells and promotes CYP1A1 and CYP3A4 mRNA expression in human hepatocytes
  • Available as a 10 mM solution in DMSO, ready for reconstitution
  • High purity of 99.49%
Catalog No. 50-004-09400