Learn More
Medchemexpress LLC Fipronil (Standard) | 120068-37-3 | 99.5% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0822R50MG
Encompass_Preferred
Fipronil (Standard) is the analytical standard of Fipronil, intended for research and analytical applications. It is a broad-spectrum insecticide effective against various pests including Lepidoptera species, thrips, locusts, ants, cockroaches, fleas, and ticks.
  • Fipronil selectively inhibits GABA receptors, with IC50s of 30 nM for cockroach GABA receptors and 1600 nM for rat GABA receptors.
  • Glutamate-gated chloride channels (GluCls), found in cockroaches but not in mammals, are sensitive to Fipronil's blocking effect.
  • It also induces apoptosis in HepG2 cells and promotes the expression of CYP1A1 and CYP3A4 mRNA in human hepatocytes.
  • This compound is an analytical standard used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • The molecular weight is 437.15, and its chemical formula is C12H4Cl2F6N4OS.
  • It appears as a white to off-white solid.
Catalog No. 50-004-07632