Learn More
TARGETMOL CHEMICALS INC FIRATEGRAST 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ02915MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Firategrast (SB 683699) is an orally active and specific (alpha)4(beta)1/(alpha)4(beta)7 integrin antagonist.Firategrast reduces the transport of lymphocytes into the central nervous system (CNS) and decreases multiple sclerosis (MS) activity.Firategrast is used in relapsing-remitting multiple sclerosis.Firategrast treatment is associated with a median cerebrospinal fluid associated with modest reductions in CD4 CD8 and CD19 lymphocyte counts. purity: 99%
Catalog No. 50-257-8800