Learn More
Essendant FIRE EXTINGUISHERSFX5.5H
Supplier:  Essendant KID21032142
Encompass_Preferred
Safeguard your property with the dependable Kidde PRO-5MP rechargeable fire extinguisher designed for commercial use to meet code compliance for larger areas and common fires involving wood paper and textiles (Class A) flammable liquids and gases (Class B) and electrical equipment (Class C) Manufactured from durable corrosion-resistant aluminum the Kidde Pro fire extinguisher is a breeze to handle it provides a remarkable discharge rate of 13 to 15 seconds a range of 12 to 18 feet and an operating pressure of 195 PSI The sleek painted aluminum handle aluminum cylinder and metal valve are impact-resistant making the Kidde Pro fire extinguisher dependable when you need it most The easy-to-pull safety pin gives you quick access for simple operation designed to combat fire hazards using the PASS (Pull Aim Squeeze Sweep) firefighting method The multi-directional discharge hose allows you to fight fire from various angles Note that the dry chemical agent in this fire extin
Catalog No. 50-000-34058