Learn More
Fireflysci Cleaning Swabs For Micro Focus Cell and Other Cuvettes, 100 pack
SDP
Supplier:  Fireflysci SWABMFC100
Encompass_Preferred
Cleaning swabs for Micro Focus Cell and other cuvettes• Comes in a pack of 100• Length of swab- 2.76 inches• Tips are made from knit polyester• Length of tip- 0.472 inches• Handle made of sealed polypropylene
Catalog No. NC2197239