Learn More
Fire King Security Group Fire Resistant - Double door storage cabinet - 72"H Platinum Finish
SDP
Supplier:  Fire King Security Group CF7236DPL
Encompass_Preferred
Double door storage cabinet. UL Class 350 fire rating and with one Medeco UL listed high-security keylock and equipped with 4 adjustable shelves. HIPAA compliance solution.
Catalog No. NC2581175
Only null left
Add to Cart
Request a Quote