Learn More
Growcells FIROZEH (X-GAL/IPTG SOLUTION)
Supplier:  Growcells MESL3001
Encompass_Preferred
Ready to use X-Gal/IPTG solution designed for selection of cloning construct in LacZ expression in the presence of lacl repressor. Stable at 4C Produced from the non-toxic ingredients without DMSO or DMF.
Catalog No. 50-103-4890