Learn More
Selleck Chemical LLC Firsocostat (GS-0976) 2mg 1434635-54-7 NDI-010976, ND-630
SDP
Supplier:  Selleck Chemical LLC S88932MG
Encompass_Preferred
Firsocostat (GS-0976, NDI-010976, ND-630) is a reversible inhibitor of acetyl CoA carboxylase (ACC) with IC50s of 2.1 nM,6.1 nM for hACC1 and hACC2,respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1902599