Learn More
American Research Products Inc Fish Estradiol E2 ELISA kit
Supplier:  American Research Products Inc CSBE13017FH
Encompass
Fish Estradiol (E2) ELISA kit from CUSABIO Detection Range 40 pg/ml-1000 pg/ml Sensitivity 40 pg/ml Sample serum plasma Method Competitive
Catalog No. 50-003-26389