Learn More
American Research Products Inc Fish free tri iodothyronine F
Supplier:  American Research Products Inc CSBEQF027510FI
Encompass
Fish free tri-iodothyronine (FT3) ELISA kit from CUSABIO Detection Range 2 pmol/L-32 pmol/L Sensitivity 0 38 pmol/L Sample serum plasma Method Competitive
Catalog No. 50-003-26415