Learn More
American Research Products Inc Fish Insulin like growth facto
Supplier:  American Research Products Inc CSBE12122FH
Encompass
Fish Insulin-like growth factor 1 IGF-1 ELISA Kit from CUSABIO Detection Range 25 pg/mL-2000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29747