Learn More
American Research Products Inc Fish lysozyme renal amyloidos
Supplier:  American Research Products Inc CSBE17296FH
Encompass
Fish lysozyme (renal amyloidosis) (LZM) ELISA kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-28504