Learn More
American Research Products Inc Fish SuperoXide Dismutase SOD
Supplier:  American Research Products Inc CSBE15929FH
Encompass
Fish SuperoXide Dismutase (SOD) ELISA kit from CUSABIO Detection Range 1 25 ng/mL-20 ng/mL Sensitivity 0 625 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26822