Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc FITC aHu CD14 Anti Primary

Supplier:  Cytek Biosciences Inc 350149T100

Encompass_Preferred

Product Name: FITC Anti-Human CD14 (61D3)
Product Class: Antibody
Type: Primary
Product Name: FITC Anti-Human CD14 (61D3)
Cytek Catalog Number: 35-0149-T100
Marker: CD14
Target Species: Human
Clonality: Monoclonal
Clone: 61D3
Host Species: Mouse
Isotype: IgG1, k
Conjugation: FITC
Size: 100 Tests

Catalog No. 50-000-85578


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.