Learn More
Bachem FITC-eAhx-Amyloid Beta Protein (1-42) Trifluoroacetate
SDP
Supplier:  Bachem 4095738.0500
Encompass_Preferred
FITC-eAhx-Amyloid Beta Protein (1-42) Trifluoroacetate, FITC-eAhx-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH, , C230H333N57O66S2, Mr 5016.65, 0.5mg. FITC-LC-Amyloid b-Protein (1-42), FITC labeled b-Amyloid (1-42).
Catalog No. 50-194-6620