Learn More
Medchemexpress LLC FITC-Ahx-Gly-Arg-Gly-Asp-Ser-Pro | 2022956-44-9 | 98.9% | 1090.12 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP48965MG
Encompass_Preferred
FITC-Ahx-Gly-Arg-Gly-Asp-Ser-Pro is a GRGDSP coupled to FITC. GRGDSP is an integrin inhibitor that can inhibit the adherence of tumor cells to endothelial cells of blood vessels and limit its metastasis. This product is for research use only and not intended for human use.
  • Integrin inhibitor
  • Synthetic linear RGD peptide
  • Fluorescent-labeled peptide
  • Used in cancer research
  • For research use only
Catalog No. 50-003-97114