Learn More
Medchemexpress LLC FITC-β-Ala-Amyloid β-Protein (1-42) (ammonium) | 99.4% | 4974.50 (free base) | 1 MG

Supplier: Medchemexpress LLC HYP39081MG
FITC-β-Ala-Amyloid β-Protein (1-42) ammonium is a fluorescently labeled Aβ1-42 peptide. This product is for research use only and plays a key role in Alzheimer's disease research.
- FITC tagged Aβ1-42 peptide
- High purity (99.36%)
- Supports Alzheimer's disease research
- Recommended storage for powder: -80°C for 2 years, -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.