Learn More
Biolegend FITC anti-CD105 (Endoglin)
Supplier:  Biolegend 800505
Encompass_Preferred
FITC anti-CD105 (Endoglin) [SN6h]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-208-9808