Learn More
Biolegend FITC anti-human CD25
Supplier:  Biolegend 302604
Encompass_Preferred
FITC anti-human CD25 [BC96]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-165-877