Learn More
Biolegend FITC anti-human CD5
Supplier:  Biolegend 300605
Encompass_Preferred
FITC anti-human CD5 [UCHT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Capuchin Monkey, Common Marmoset, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-165-606