Learn More
Biolegend FITC anti-human CD81 (TAPA-1)
Supplier:  Biolegend 349504
Encompass_Preferred
FITC anti-human CD81 (TAPA-1) [5A6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-168-525